Hyaluronic acid high sulphated
Catalog # TCIAH1739-10MG
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Content of sulfur:13.0 to 15.5 %
- Drying loss:max. 20 %
Specification Test Results
About this item
:
:776.651 g/mol
Storage Temperature:Refrigerator
CAS Number:9004-61-9
Formula:C₂₈H₄₄N₂O₂₃
MDL Number:MFCD00131348