Order Entry
Finland
ContactUsLinkComponent
[4-[(2-Hydroxytetradecyl)oxy]phenyl]phenyliodonium Hexafluoroantimonate ≥98.0% (by HPLC)
Toxic
Environment
[4-[(2-Hydroxytetradecyl)oxy]phenyl]phenyliodonium Hexafluoroantimonate ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
Toxic
Environment
[4-[(2-Hydroxytetradecyl)oxy]phenyl]phenyliodonium Hexafluoroantimonate ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

Frequently Bought Together