Hyaluronate Dodecasaccharide ≥95.0% (by HPLC)
Catalog # TCIAH1666-1MG
Supplier: TCI
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- TOF-MS:to pass test
- NMR:to pass test
Specification Test Results
About this item
:
:2293.92 g/mol
Storage Temperature:Freezer
CAS Number:71058-16-7
Formula:C₈₄H₁₂₈N₆O₆₇