
2-Hydroxypyrazine ≥98.0% (by GC, titration analysis)
Catalog # TCIAH1580-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:185.0 to 189.0 deg-C
Specification Test Results
About this item
:
:96.09 g/mol
EINECS:228-455-2
:GHS07
CAS Number:6270-63-9
:MFCD11877813
Formula:C₄H₄N₂O
MDL Number:MFCD00233966