
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:55959-84-7
:
Formula:C₃H₉BrN₄
MDL Number:MFCD00012694
:181.04 g/mol
EINECS:259-921-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Potassium iodate Method):min. 98.0 %
- Melting point:183.0 to 187.0 deg-C
Specification Test Results
Frequently Bought Together