
2'-Hydroxy-5'-methylacetanilide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:6375-17-3
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00020183
:165.19 g/mol
EINECS:228-934-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:158.0 to 162.0 deg-C
Specification Test Results
Frequently Bought Together