
1-Hexylpyridinium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:797789-00-5
:
Formula:C₁₁H₁₈F₆NP
MDL Number:MFCD03093298
:309.23 g/mol
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
- Melting point:46.0 to 50.0 deg-C