Order Entry
Finland
ContactUsLinkComponent
 
Irritant
5-Hydroxy-2-(trifluoromethyl)pyridine ≥98.0% (by GC)
Catalog # TCIAH1406-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
5-Hydroxy-2-(trifluoromethyl)pyridine ≥98.0% (by GC)
Catalog # TCIAH1406-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :163.1 g/mol
 :GHS07
CAS Number:216766-12-0
Formula:C₆H₄F₃NO
MDL Number:MFCD06801325