Order Entry
Finland
ContactUsLinkComponent
7-Hydroxy-4-methylcoumarin-3-acetic acid ≥97.0% (by HPLC, titration analysis)
7-Hydroxy-4-methylcoumarin-3-acetic acid ≥97.0% (by HPLC, titration analysis)
  TCIAH1398-100MG
 :  TCI
 :  
undefined
7-Hydroxy-4-methylcoumarin-3-acetic acid ≥97.0% (by HPLC, titration analysis)
  TCIAH1398-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %

 

 

 :
 :234.21 g/mol
 :268°C
 :GHS07
 :Freezer
 :5852-10-8
 :C₁₂H₁₀O₅
 :MFCD00467589