
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:106627-54-7
:
Formula:C₄H₄NNaO₆S
MDL Number:MFCD00043100
:217.13 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %