
4-Hydroxy-2-methoxybenzaldehyde ≥97.0% (by GC, titration analysis)
: TCI
:18278-34-7
:
:C₈H₈O₃
:MFCD00051964
:152.15 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:156.0 to 160.0 deg-C
Frequently Bought Together