Hyaluronate Octasaccharide ≥95.0% (by HPLC)
TCIAH1148-1MG
: TCI
:
- Pk:1 mg
- Seq:2070
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- TOF-MS:to pass test
- NMR:to pass test
:
:1535.29 g/mol
:Freezer
:57323-43-0
:C₅₆H₈₆N₄O₄₅