4-(4-Hydroxyphenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
Catalog # TCIAH1063-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:154.0 to 158.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:227.28 g/mol
CAS Number:103661-13-8
Formula:C₁₀H₁₃NO₃S
MDL Number:MFCD06797109