1,2-Hexadecanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6920-24-7
:
Formula:C₁₆H₃₄O₂
MDL Number:MFCD00004727
:258.45 g/mol
EINECS:230-030-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:74.0 to 78.0 deg-C