3-Hydroxy-2-naphthoic acid-2-chloroanilide ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0783-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:228.0 to 232.0 deg-C
- Solubility in N,N-DMF:almost transparency
Specification Test Results
About this item
:
:297.74 g/mol
CAS Number:6704-40-1
Formula:C₁₇H₁₂ClNO₂
MDL Number:MFCD00191652