Potassium Hyaluronate (from cockscomb)
Catalog # TCIAH0652-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Drying loss:max. 20.0 %
- Molecular weight:min. 8000 (calcd.on dried substance)
Specification Test Results
About this item
:
CAS Number:31799-91-4
MDL Number:MFCD00131349