Hyaluronic acid sodium salt (from cockscomb)
Supplier: TCI
Certificates
About this item
CAS Number:9067-32-7
:
Formula:(C₁₄H₂₁NaNO₁₁)n
MDL Number:MFCD00875848
Merck Index:14,04757
Specification Test Results
- Appearance:White powder to crystal
- Molecular weight:min. 350,000 (calcd.on dried substance)
- Drying loss:max. 20 %