Hyaluronic acid (from cockscomb)
Catalog # TCIAH0595-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Specific rotation [a]20/D:-60.0 to -73.0 deg (C=0.1, H2O) (calcd.on dried substance)
- Drying loss:max. 12.0 %
- Molecular weight:5000 to 150000 (calcd.on dried substance)
Specification Test Results
About this item
:
Merck Index:14,04757
:776.651 g/mol
EINECS:232-678-0
CAS Number:9004-61-9
Formula:C₂₈H₄₄N₂O₂₃
MDL Number:MFCD00131348