

1,5-Heptadiene (cis and trans mixture) cis- and trans-mixture ≥97.0% (by GC)
Catalog # TCIAH0551-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:96.17 g/mol
Flash Pt:-3°C
UN:3295
ADR:3,II
:GHS02
:GHS08
CAS Number:1541-23-7
Formula:C₇H₁₂
MDL Number:MFCD00026998
Flash Pt:–3°C