
1-(2-Hydroxyethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2955-88-6
:
Formula:C₆H₁₃NO
MDL Number:MFCD00003181
:115.18 g/mol
Flash Pt:56°C
EINECS:220-976-3
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %