Hemin chloride (from porcine) ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16009-13-5
:
Formula:C₃₄H₃₂ClFeN₄O₄
MDL Number:MFCD00010726
:651.95 g/mol
Merck Index:14,04644
EINECS:240-140-1
Specification Test Results
- Appearance:Gray to Dark blue to Black powder to crystaline
- Purity (Chelometric Titration):min. 95.0 %
- Molar extinction coefficient:min. 54000 (pH 10.0, Carbonate Buffer, 383.0 to 387.0 nm)
- Drying loss:max. 1.0 %