Order Entry
Finland
ContactUsLinkComponent
prim-O-Glucosylcimifugin ≥97.0% (by HPLC)
prim-O-Glucosylcimifugin ≥97.0% (by HPLC)
Supplier:  TCI
Certificates
prim-O-Glucosylcimifugin ≥97.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Specific rotation [a]20/D:
    14.0 to +18.0 deg (C=0.3, MeOH)

Specification Test Results