
2-[(Prop-2-ynyloxy)methyl]oxirane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18180-30-8
:
Formula:C₆H₈O₂
MDL Number:MFCD00099300
:112.13 g/mol
Boiling Pt:95°C
Flash Pt:69°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %