Order Entry
Finland
ContactUsLinkComponent
 
2-O-ɑ-D-glucopyranosyl-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
2-O-ɑ-D-glucopyranosyl-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +188 to +198 deg (C=1, H2O)