Glycyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAG0190-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-1.0 to +1.0 deg (C=2, 5mol/L HCL)
Specification Test Results
About this item
:
:261.28 g/mol
EINECS:218-581-6
CAS Number:2189-26-6
Formula:C₁₃H₁₅N₃O₃
MDL Number:MFCD00022767