Monostearin >60.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:123-94-4
:
Formula:C₂₁H₄₂O₄
MDL Number:MFCD00036186
:358.56 g/mol
Merck Index:14,04489
EINECS:204-664-4
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 60.0 % (total of isomer)
- Melting point:68.0 to 72.0 deg-C