
3-Fluoro-2-methoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:856250-60-7
:
Formula:C₆H₇BFNO₃
MDL Number:MFCD04112520
:170.9341 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 110.0 %
Specification Test Results
Frequently Bought Together