


Fenpyroximate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:134098-61-6
:
Formula:C₂₄H₂₇N₃O₄
MDL Number:MFCD00274596
:421.50 g/mol
Merck Index:14,03996
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):9.50 to 10.50 %
- Melting point:100.0 to 104.0 deg-C