2-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:452972-14-4
:
Formula:C₁₁H₁₅BFNO₂
MDL Number:MFCD06798245
:223.05 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:45.0 to 49.0 deg-C