
2-Fluoro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:444120-95-0
:
Formula:C₁₁H₁₅BFNO₂
MDL Number:MFCD03412792
:223.05 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:29.0 to 33.0 deg-C
Specification Test Results
Frequently Bought Together