
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:481681-02-1
:
Formula:C₇H₈BFO₃
MDL Number:MFCD08235075
:169.95 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):97.0 to 112.0 %