2-Fluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
: TCI
:876062-39-4
:
:C₁₂H₁₆BFO₂
:MFCD05663874
:222.07 g/mol
- Appearance:White to Almost white clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %