Order Entry
Finland
ContactUsLinkComponent
 
Fmoc-Lys(biotin)-OH ≥95.0% (by HPLC, titration analysis)
 :  TCI
 
Fmoc-Lys(biotin)-OH ≥95.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Neutralization titration):
    min. 95.0 %
  • Specific rotation [a]20/D:
    +18.0 to +21.0 deg (C=1,DMSO)

 

Frequently Bought Together