Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-3-nitrobenzonitrile
4-Fluoro-3-nitrobenzonitrile ≥98.0% (by GC)
4-Fluoro-3-nitrobenzonitrile ≥98.0% (by GC)
:
TCI
4-Fluoro-3-nitrobenzonitrile ≥98.0% (by GC)
:
TCI
:
1009-35-4
:
:
C₇H₃FN₂O₂
:
MFCD01632197
:
166.11 g/mol
:
3439
:
6.1,III
:
GHS06
Appearance:
White to Yellow to Green powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
91.0 to 95.0 deg-C