1-Fluoro-3,3-dimethyl-1,2-benziodoxole ≥98.0% (by titrimetric analysis)
Catalog # TCIAF0957-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:86.0 to 90.0 deg-C
Specification Test Results
About this item
:
:280.08 g/mol
:GHS07
CAS Number:1391728-13-4
Formula:C₉H₁₀FIO
MDL Number:MFCD28397076