
3-Fluoro-2-nitrophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:385-01-3
:
Formula:C₆H₄FNO₃
MDL Number:MFCD07368753
:157.10 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:36.0 to 40.0 deg-C
Specification Test Results
Frequently Bought Together