Order Entry
Finland
ContactUsLinkComponent
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥98.0% (by GC)
Irritant
Environment
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
Environment
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    129.0 to 133.0 deg-C

Specification Test Results