Flavoxate hydrochloride ≥98.0% (by HPLC, titration analysis)
: TCI
:3717-88-2
:
:C₂₄H₂₅NO₄
:MFCD00072099
:427.93 g/mol
:234°C
:223-066-4
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %