Order Entry
Finland
ContactUsLinkComponent
 
Neomycin sulphate ≥97.0% (by total nitrogen basis)
  TCIAF0649-25G
 :  TCI
 :  
Neomycin sulphate ≥97.0% (by total nitrogen basis)
  TCIAF0649-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 % (calcd.on dried substance)
  • Specific rotation [a]20/D:
    +53.0 to +60.0 deg (C=10, H2O) (calcd.on dried substance)
  • Drying loss:
    max. 8.0 %

 

 

 :
 :14,06454
 :908.87 g/mol
 :215-773-1
 :GHS07
 :1405-10-3
 :1404-04-2
 :C₂₃H₄₆N₆O₁₃·3H₂SO₄
 :MFCD00077833