Order Entry
Finland
ContactUsLinkComponent
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Specific rotation [a]20/D:
    +17.0 to +21.0 deg (C=1, DMF)
  • Water:
    max. 7.0 %

Specification Test Results

Related Information

Frequently Bought Together