Nɑ-Formyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF0134-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:232.24 g/mol
CAS Number:16108-03-5
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD00055983