
Ethylene Glycol bis(2-aminophenyl) ether ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52411-34-4
:
Formula:C₁₄H₁₆N₂O₂
MDL Number:MFCD00087648
:244.29 g/mol
Boiling Pt:225°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:131.0 to 135.0 deg-C