Order Entry
Finland
ContactUsLinkComponent
Ethidium bromide 0.5mg/mL in water for electrophoresis
Ethidium bromide 0.5mg/mL in water for electrophoresis
Catalog # TCIAE1363-10ML
Supplier:  TCI
CAS Number:  
undefined
Ethidium bromide 0.5mg/mL in water for electrophoresis
Catalog # TCIAE1363-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • DNA staining test:
    to pass test

Specification Test Results

About this item

 :
Merck Index:14,04731
 :Packaged in dropper bottle.
 :394.31 g/mol
Storage Temperature:Refrigerator
EINECS:214-984-6
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724