
9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:856422-41-8
:
Formula:C₂₆H₃₆BNO₂
MDL Number:MFCD31618140
:405.39 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
Specification Test Results
Frequently Bought Together