Epmedin C ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:110642-44-9
:
Formula:C₃₉H₅₀O₁₉
MDL Number:MFCD04039820
:822.81 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Specific rotation [a]20/D:-103 to -113 deg (C=0.2, MeOH)