
Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:16078-71-0
:
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD00020731
:231.26 g/mol
EINECS:240-224-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:98.0 to 102.0 deg-C