
trans,trans-4'-Ethylbicyclohexyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAE1251-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Mesomorphic range:178.0 to 233.0 deg-C
Specification Test Results
About this item
:
:238.37 g/mol
:GHS07
CAS Number:84976-67-0
Formula:C₁₅H₂₆O₂
MDL Number:MFCD05864406