
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:50920-65-5
:
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD00156147
:154.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:142.0 to 146.0 deg-C
Specification Test Results
Frequently Bought Together