
Ethyl-4-chloroquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13720-94-0
:
Formula:C₁₂H₁₀ClNO₂
MDL Number:MFCD00173407
Boiling Pt:129°C
:235.67 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:44.0 to 48.0 deg-C
Specification Test Results
Frequently Bought Together