trans,trans-4'-Ethyl-4-(4-trifluoromethoxyphenyl)bicyclohexyl ≥98.0% (by GC)
Catalog # TCIAE1165-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
- Mesomorphic range:72.0 to 121.0 deg-C
Specification Test Results
About this item
:
:354.46 g/mol
CAS Number:135734-59-7
Formula:C₂₁H₂₉F₃O
MDL Number:MFCD09842837